Learn More
Chemglass Life Sciences Reaction Vessel Lid, 5-Neck, 200mm
SDP
Catalog No. CG194650
Encompass_Preferred
Reaction Vessel Lid, 5-Neck, 200mm. 5-Neck reaction vessel lid for 200mm flanged vessels. Lid has a CN 45/50, 3-SN 45/50, and 1-SN 60mm Duran o-ring flange angled for powder addition.PLEASE NOTE: 60MM LID, O-RING AND CLAMP MUST BE ORDERED SEPARATELY.
Catalog No. CG194650 Supplier Chemglass Life Sciences Supplier No. CG194650
Reaction Vessel Lid, 5-Neck, 200mm. 5-Neck reaction vessel lid for 200mm flanged vessels. Lid has a CN 45/50, 3-SN 45/50, and 1-SN 60mm Duran o-ring flange angled for powder addition.PLEASE NOTE: 60MM LID, O-RING AND CLAMP MUST BE ORDERED SEPARATELY.