Learn More
Agilent Technologies 200MU/80UL GLYKO N-GLYCANASE
Catalog No. NC9046465
Encompass
PNGase F, peptide-N-glycosidase F,peptide-N4-(N-acetyl--glucosaminyl)asparagine amidase.Releases intact N-glycans by cleavingbetween the innermost GlcNAc and Asn.
Catalog No. NC9046465 Supplier Agilent Technologies Supplier No. GKE5006B
PNGase F, peptide-N-glycosidase F,peptide-N4-(N-acetyl--glucosaminyl)asparagine amidase.Releases intact N-glycans by cleavingbetween the innermost GlcNAc and Asn.