Learn More
Hach Company 200 mV ORP Solution, 1L
Catalog No. 501997407 Shop All Hach Company Products
Encompass
ORP Reference Solution used for ORP sensor calibration (in resealable plastic bottles).
Catalog No. 50-199-7407 Supplier Hach Company Supplier No. 25M2A1001119
ORP Reference Solution used for ORP sensor calibration (in resealable plastic bottles).