Learn More
Malvern Panalytical 200 nm Latex Transfer Standards 3 mL
Catalog No. NC1379642
Encompass
200nm size calibrated Standards, highly uniform polystyrene spheres, packaged as aqueous suspensions.
Catalog No. NC1379642 Supplier Malvern Panalytical Supplier No. 10042974
200nm size calibrated Standards, highly uniform polystyrene spheres, packaged as aqueous suspensions.