Learn More
CELLTREAT Scientific Products 200L/300L Pipette Tip Rack, Empty Rack for Reload System, Non-sterile
SDP
Supplier:  CELLTREAT Scientific Products 229061
Encompass_Preferred
Hinged lids on rack are designed for easy one-handed opening, making it easier to access repeatedly. Lids have stacking feature for added stability and a secure locking tab. Lightweight lid design prevents tipping when open. Interior support ribs prevent racks from warping. Empty rack without wafer allows refill from pipette tip reload systems. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed non-sterile, 20/case.
Catalog No. 50-165-6734