Learn More
Thomas Scientific TIPS, 200UL BARRIER, STERILE, MULTIGRIP, 10X96/RACK
SDP
Supplier:  Thomas Scientific 1158T50PK
Encompass
MultiGrip™ pipet tips from Denville Scientific are the best tips for multichannel pipettors. This revolutionary tip is manufactured from two different resins which requires only 1/10 the sealing pressure of other leading pipet tips and reduces wear on pipettors. The soft seal offers a more universal fit, which standardizes most popular multichannel pipettors on one line of tips. MultiGrip™… The Multichannel Solution!
Catalog No. NC1642371