Learn More
CELLTREAT Scientific Products 200 ul Filter Pippette Tips, 960/case
SDP
Supplier:  CELLTREAT Scientific Products 229072
Encompass_Preferred
Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tips. Pipette tips are packaged in racks with hinged lids designed for easy one-handed opening. 10μL tips feature an extended length for easier access to the bottom of vessels. Fine tip for improved accuracy and precision. Filter tips are fitted with a polymer barrier to reduce the risk of contamination and prevent aerosols from penetrating into the pipette. Lids feature stacking supports, and are removable to save bench space.
Catalog No. NC2206444