Learn More
USA Scientific Inc 200ul flat gel tip, .37 mm
Catalog No. 502744917
Encompass
200 uL flat sequencing gel tip, 0.37 mm OD orifice, 96 tips/rack, 2 racks/pack (192 tips)
Catalog No. 50-274-4917 Supplier USA Scientific Inc Supplier No. 10224600
200 uL flat sequencing gel tip, 0.37 mm OD orifice, 96 tips/rack, 2 racks/pack (192 tips)