Learn More
Agilent Technologies 200UL GLYKO SIALIDASE A/NANASE
Catalog No. NC9067497
Encompass
Releases(2-3)-, (2-6)-, (2-8), and (2-9)-linked N-acetylneuraminic acid fromoligosaccharides and glycoproteins.Also capable of releasing N-glycolylneuraminic acid.
Catalog No. NC9067497 Supplier Agilent Technologies Supplier No. GK80040
Releases(2-3)-, (2-6)-, (2-8), and (2-9)-linked N-acetylneuraminic acid fromoligosaccharides and glycoproteins.Also capable of releasing N-glycolylneuraminic acid.