Learn More
Ops Diagnostics 200 micron Low Binding Zirconium Beads, 250 gm
Catalog No. NC1280038
Encompass
Low Binding Beads are chemically treated so they bind less solutes liberated from homogenized samples.  Untreated silica and zirconium beads have surface chemistries that bind DNA, RNA, and proteins upon disruption of cells.  Both enzyme and PCR-based assays have lower sensitivity when analytes are liberated using untreated beads.  Low Binding Beads yield a greater concentration of solutes and provide greater range and linearity, especially in real-time PCR assays.
Catalog No. NC1280038 Supplier Ops Diagnostics Supplier No. BLBZ 20025009
Low Binding Beads are chemically treated so they bind less solutes liberated from homogenized samples.  Untreated silica and zirconium beads have surface chemistries that bind DNA, RNA, and proteins upon disruption of cells.  Both enzyme and PCR-based assays have lower sensitivity when analytes are liberated using untreated beads.  Low Binding Beads yield a greater concentration of solutes and provide greater range and linearity, especially in real-time PCR assays.