Learn More
CELLTREAT Scientific Products 200L Low Retention Pipette Tip Reload System, Non-sterile
SDP
Catalog No. 501656730
Encompass_Preferred
Reload System offers clear window for easy identification of color-coded wafers. Support wafers have large grasping tabs for easy removal from Reload System and Rack. Secure locking tab keeps reload system closed and prevents tumbling tips. Reduce plastic waste and save time by refilling empty racks.
Catalog No. 50-165-6730 Supplier CELLTREAT Scientific Products Supplier No. 229054
Reload System offers clear window for easy identification of color-coded wafers. Support wafers have large grasping tabs for easy removal from Reload System and Rack. Secure locking tab keeps reload system closed and prevents tumbling tips. Reduce plastic waste and save time by refilling empty racks.