Learn More
USA Scientific Inc LTS® compatible tips for Rainin® pipettes. Low retention. 200ul, Filter rack
Catalog No. NC2889198
Encompass
These LTS® Compatible Tips are designed to perfectly fit both single- and multi-channel Rainin® pipettes ensuring precise and accurate sample volume delivery. Made from virgin polypropylene and designed using a hydrophobic resin technology that enables low liquid retention and high sample recovery. The refill wafers are designed to fit only in the USA Scientific LTS® Compatible Tip racks, which are available separately with the following part numbers: 1070-1901 (20-200 μL), and 1070-3901 (1000 µl XL). Racks come in 10 racks (960 tips.)LTS® and Rainin® are registered trademarks of METTLER-TOLEDO GMBH (Switzerland).
Catalog No. NC2889198 Supplier USA Scientific Inc Supplier No. 10711811
These LTS® Compatible Tips are designed to perfectly fit both single- and multi-channel Rainin® pipettes ensuring precise and accurate sample volume delivery. Made from virgin polypropylene and designed using a hydrophobic resin technology that enables low liquid retention and high sample recovery. The refill wafers are designed to fit only in the USA Scientific LTS® Compatible Tip racks, which are available separately with the following part numbers: 1070-1901 (20-200 μL), and 1070-3901 (1000 µl XL). Racks come in 10 racks (960 tips.)LTS® and Rainin® are registered trademarks of METTLER-TOLEDO GMBH (Switzerland).