Learn More
USA Scientific Inc 200 ul TipOne RPT tip starter system
Catalog No. 507541537
Encompass
TipOne RPT 200 ul ultra low retention beveled pipet tip starter system, 10 racks and 2 packs of refill wafers (2880 tips)
Catalog No. 50-754-1537 Supplier USA Scientific Inc Supplier No. 11611500
TipOne RPT 200 ul ultra low retention beveled pipet tip starter system, 10 racks and 2 packs of refill wafers (2880 tips)