Learn More
Genesee Scientific Corporation Genesee Scientific 200 µL Olympus Barrier Tips, Low Binding, Sterile, Universal Fit
SDP
Catalog No. NC1962414
Encompass
The Genesee Scientific 200 µL Olympus Barrier Tips are premium low-binding, filtered pipette tips designed for precise liquid handling in scientific applications. Made from virgin polypropylene and pre-sterilized by gamma irradiation, these tips ensure minimal sample retention and enhance recovery. Universal fit compatibility with most research-grade pipettors distinguishes them, promising the integrity vital for analytical precision. Their racked packaging further simplifies lab use.
Catalog No. NC1962414 Supplier Genesee Scientific Corporation Supplier No. 26412
The Genesee Scientific 200 µL Olympus Barrier Tips are premium low-binding, filtered pipette tips designed for precise liquid handling in scientific applications. Made from virgin polypropylene and pre-sterilized by gamma irradiation, these tips ensure minimal sample retention and enhance recovery. Universal fit compatibility with most research-grade pipettors distinguishes them, promising the integrity vital for analytical precision. Their racked packaging further simplifies lab use.