Learn More
Alkali Scientific Alkali Scientific™ 200uL PerkinElmer (Revvity) Janus™ MultiProbe Compatible Black Conductive Robotic Automation Tips (P200), 96-Well Head/Tip Format, Low Retention, Sterile - 960/Box, 4,800/Case, Non-Filtered
SDP
Catalog No. 503028965
Encompass_Preferred
Alkali Scientific™  PerkinElmer-compatible automation tips  are compatible with the Fontus and the JANUS G3 VariSpan™ pipetting module and can be used for a variety of automated liquid handling applications. Each individual tip has a small amount of graphite added to the polypropylene in the manufacturing process to enable an electric current in the tips. This allows the VariSpan pipetting module to detect the surface of liquids and pipette from a defined depth relative to the surface of the liquid. Specifications: Working Volume: 2.00 – 200.00 µL Well Format: 96 | 96, 384 plate accessible Head Compatibility: Janus Varispan
Catalog No. 50-302-8965 Supplier Alkali Scientific Supplier No. PEBST1200
Alkali Scientific™  PerkinElmer-compatible automation tips  are compatible with the Fontus and the JANUS G3 VariSpan™ pipetting module and can be used for a variety of automated liquid handling applications. Each individual tip has a small amount of graphite added to the polypropylene in the manufacturing process to enable an electric current in the tips. This allows the VariSpan pipetting module to detect the surface of liquids and pipette from a defined depth relative to the surface of the liquid. Specifications: Working Volume: 2.00 – 200.00 µL Well Format: 96 | 96, 384 plate accessible Head Compatibility: Janus Varispan