Learn More
Alkali Scientific 200uL PerkinElmer Revvity Janu
SDP
Catalog No. 503029033
Encompass_Preferred
Alkali Scientifichas designed its PerkinElmer automation tips for seamless compatibility with PerkinElmer Janus Varispan liquid handlers When using our PerkinElmer Janus-compatible tips theres no need for a separate labware definition and PerkinElmer Janus automation control software protocols remain unchanged This 96-format conductive black tip is fully interchangeable with the original PerkinElmer P200 automation pipette tip These tips are also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsThe PerkinElmer P200 conductive black tip is designed for volume transfers up to 200 L offering outstanding precision and accuracy Its capacity and length provide flexibility for protocol design making it versatile for a range of applications These tips are compatible with both 96-format and 384-format plates and fit Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P200 tips using pre
Catalog No. 50-302-9033 Supplier Alkali Scientific Supplier No. PEST1200
Alkali Scientifichas designed its PerkinElmer automation tips for seamless compatibility with PerkinElmer Janus Varispan liquid handlers When using our PerkinElmer Janus-compatible tips theres no need for a separate labware definition and PerkinElmer Janus automation control software protocols remain unchanged This 96-format conductive black tip is fully interchangeable with the original PerkinElmer P200 automation pipette tip These tips are also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsThe PerkinElmer P200 conductive black tip is designed for volume transfers up to 200 L offering outstanding precision and accuracy Its capacity and length provide flexibility for protocol design making it versatile for a range of applications These tips are compatible with both 96-format and 384-format plates and fit Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P200 tips using pre