Learn More
Bellco Glass, Inc. 200µL Pipette, MLA, Precision, Red
SDP
Catalog No. 502329336
Encompass_Preferred
200µL Pipette, MLA, Precision, RedTHE CLASSIC MODELSingle-stroke “to deliver” operationAvailable in 28 fixed volumes ranging from 5 μL- 1000 μLEasily adjusted as needed to achieve accurate performance on dense or viscous liquids All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorRedPrimary MaterialAluminum Alloy & Stainless SteelVolume200 μLVistaLab Tip Fit SizeSmall 250μLUniversal FitNoAccuracy±1.00%Precision0.30%
Catalog No. 50-232-9336 Supplier Bellco Glass, Inc. Supplier No. 451900006
200µL Pipette, MLA, Precision, RedTHE CLASSIC MODELSingle-stroke “to deliver” operationAvailable in 28 fixed volumes ranging from 5 μL- 1000 μLEasily adjusted as needed to achieve accurate performance on dense or viscous liquids All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorRedPrimary MaterialAluminum Alloy & Stainless SteelVolume200 μLVistaLab Tip Fit SizeSmall 250μLUniversal FitNoAccuracy±1.00%Precision0.30%