Learn More
CELLTREAT Scientific Products 200L Pipette Tips, Bulk Packed, Non-sterile
SDP
Catalog No. 501656726
Encompass_Preferred
Pipette Tips deliver high performance with exceptional value. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. 200¿L filter tip is longer than standard tips for better access to vessels & increased headspace to filter. Molded volume reference markings offer a quick double check opportunity. Bulk, non-sterile option available in unique self-standing, resealable bags. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed bulk non-sterile, 1000/re-sealable bag, 5000/case.
Catalog No. 50-165-6726 Supplier CELLTREAT Scientific Products Supplier No. 229044
Pipette Tips deliver high performance with exceptional value. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. 200¿L filter tip is longer than standard tips for better access to vessels & increased headspace to filter. Molded volume reference markings offer a quick double check opportunity. Bulk, non-sterile option available in unique self-standing, resealable bags. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed bulk non-sterile, 1000/re-sealable bag, 5000/case.