Learn More
Bellco Glass, Inc. 200µL Pipette Tips, Bulk
SDP
Catalog No. 502329244
Encompass_Preferred
Bulk, non-sterile Pipette Tips available in unique self-standing, resealable bagsPipette Tips are universal fit and compatible with major pipettesPipette Tips have smooth surfaces to ensure accurate pipettingMolded volume reference markings offer a quick double check opportunityEmpty racks are available with a wafer to allow refill from bulk packed pipette tips Pack Description1000/Re-sealable BagCase Quantity5000SterileNoRNase/DNase FreeYesAutoclavableYesLow RetentionNoFilterNoRackedNoPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume200µLGraduatedYes
Catalog No. 50-232-9244 Supplier Bellco Glass, Inc. Supplier No. 229044000
Bulk, non-sterile Pipette Tips available in unique self-standing, resealable bagsPipette Tips are universal fit and compatible with major pipettesPipette Tips have smooth surfaces to ensure accurate pipettingMolded volume reference markings offer a quick double check opportunityEmpty racks are available with a wafer to allow refill from bulk packed pipette tips Pack Description1000/Re-sealable BagCase Quantity5000SterileNoRNase/DNase FreeYesAutoclavableYesLow RetentionNoFilterNoRackedNoPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume200µLGraduatedYes