Learn More
USA Scientific Inc 200ul round gel tip, sterile
Catalog No. 502745393
Encompass
200 uL Gel-Saver round gel loading tip, 0.6 mm OD orifice, sterilized, 5 racks of 204 (1020 tips)
Catalog No. 50-274-5393 Supplier USA Scientific Inc Supplier No. 10220610
200 uL Gel-Saver round gel loading tip, 0.6 mm OD orifice, sterilized, 5 racks of 204 (1020 tips)