Learn More
Andwin Scientific 200UL STERILE TIPS HINGE BOX
SDP
Catalog No. NC9445780
Encompass_Preferred
The Signature™ Bevel Point Pipet Tips offer maximum sample recovery and minimal fluid retention. This product features compatibility most pipettors. The tips are autoclavable up to 122C (252F) and its beveled ends provide better accuracy and reprodu
Catalog No. NC9445780 Supplier Andwin Scientific Supplier No. 53508794
The Signature™ Bevel Point Pipet Tips offer maximum sample recovery and minimal fluid retention. This product features compatibility most pipettors. The tips are autoclavable up to 122C (252F) and its beveled ends provide better accuracy and reprodu