Learn More
Genesee Scientific Corporation Genesee Scientific 200 µL Sterile Filtered Pipette Tips, Low Binding, RNase-Free, 4800 Tips/Case
SDP
Catalog No. NC1909001
Encompass
Genesee Scientific 200 µL Sterile Filtered Pipette Tips are designed for precise liquid handling in scientific applications. Their low-binding surface minimizes sample retention, enhancing accuracy and recovery. Made from virgin polypropylene and pre-sterilized, they are RNase, DNase, and pyrogen-free, ensuring reliable performance for sensitive assays. The universal fit design accommodates most pipettors and comes in convenient racks, setting them apart from other products.
Catalog No. NC1909001 Supplier Genesee Scientific Corporation Supplier No. 24412C
Genesee Scientific 200 µL Sterile Filtered Pipette Tips are designed for precise liquid handling in scientific applications. Their low-binding surface minimizes sample retention, enhancing accuracy and recovery. Made from virgin polypropylene and pre-sterilized, they are RNase, DNase, and pyrogen-free, ensuring reliable performance for sensitive assays. The universal fit design accommodates most pipettors and comes in convenient racks, setting them apart from other products.