Learn More
Alkali Scientific 200uL Tecan MCA Robotic Automa
SDP
Catalog No. 503027681
Encompass_Preferred
Alkali Scientifichas designed its Tecan P200 MCA (MultiChannel Arm) tips to ensure optimal compatibility with Fluent robotic liquid handlers When using our Fluent MCA-compatible automation tips there is no need for a separate labware definition and Tecan automation control software protocols remain unchanged This 96-format tip is fully interchangeable with the original Tecan P200 automation pipette tip This 96-format tip is designed for large volume transfers up to 200 L (with filter) The Tecan P200 tip was specifically developed to offer additional volume without altering its length providing greater versatility in protocol design These tips can access both 96-format and 384-format plates and are compatible with all 96 MCA heads on both Freedom EVO and Fluent liquid handlers Alkali Scientificmanufactures its Tecan Freedom EVO and Fluent-compatible P200 tips from premium virgin polypropylene We utilize advanced polishing techniques to ensure low liquid retention without the use
Catalog No. 50-302-7681 Supplier Alkali Scientific Supplier No. MCAST0200
Alkali Scientifichas designed its Tecan P200 MCA (MultiChannel Arm) tips to ensure optimal compatibility with Fluent robotic liquid handlers When using our Fluent MCA-compatible automation tips there is no need for a separate labware definition and Tecan automation control software protocols remain unchanged This 96-format tip is fully interchangeable with the original Tecan P200 automation pipette tip This 96-format tip is designed for large volume transfers up to 200 L (with filter) The Tecan P200 tip was specifically developed to offer additional volume without altering its length providing greater versatility in protocol design These tips can access both 96-format and 384-format plates and are compatible with all 96 MCA heads on both Freedom EVO and Fluent liquid handlers Alkali Scientificmanufactures its Tecan Freedom EVO and Fluent-compatible P200 tips from premium virgin polypropylene We utilize advanced polishing techniques to ensure low liquid retention without the use