Learn More
Labcon North America Eclipse™ 200uL Graduated Pipet Tips, in 192 Stack Racks
Catalog No. NC0829622
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Graduated; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Clear; 192 tips per rack; 5 racks per stack (includes 1 free refillable rack); 10 stacks per case; 49.7mm. H; Made in USA
Catalog No. NC0829622 Supplier Labcon North America Supplier No. 10937000189
Tip, Pipet; Labcon; Eclipse™; Graduated; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Clear; 192 tips per rack; 5 racks per stack (includes 1 free refillable rack); 10 stacks per case; 49.7mm. H; Made in USA