Learn More
Dot Scientific 200UL TIP (960/PK)
SDP
Catalog No. NC9923343
Encompass
Premium Fit Filter Tip 20-200ul Beveled Graduated Natural Sterile 96 Tips Per Rack 960 Tips Per Pack
Catalog No. NC9923343 Supplier Dot Scientific Supplier No. UG2812RS
Premium Fit Filter Tip 20-200ul Beveled Graduated Natural Sterile 96 Tips Per Rack 960 Tips Per Pack