Learn More
Alkali Scientific 200uL Zymark-Style Clear Robot
SDP
Catalog No. 503026389
Encompass_Preferred
Our robotic pipette tips are meticulously crafted to meet the exact specifications of original equipment manufacturer (OEM) tips ensuring seamless compatibility and a secure fit with the industrys most popular liquid handling systems These tips are designed for use with Zymark RapidPlate Zymark SciClone Tecan Te-MO Tecan Genmate and Tecan Aquariusautomated liquid handling platforms Precision-engineered to match OEM dimensions they guarantee flawless automation performance and form a tight reliable seal with the robotic system Made from a specialized polymer that resists DNA RNA and protein adherence these tips are manufactured using diamond-polished molds to eliminate cavities or occlusions that could cause sample retention They are also free from secondary substances such as silicone ensuring sample purity Specifications Volume 200 L Color Clear Sterile Standard Style Certification RNase DNase and Pyrogen-Free Packaging 96 Tips/Rack 10 Racks/Unit 5 Units/Case
Catalog No. 50-302-6389 Supplier Alkali Scientific Supplier No. ZFT1200
Our robotic pipette tips are meticulously crafted to meet the exact specifications of original equipment manufacturer (OEM) tips ensuring seamless compatibility and a secure fit with the industrys most popular liquid handling systems These tips are designed for use with Zymark RapidPlate Zymark SciClone Tecan Te-MO Tecan Genmate and Tecan Aquariusautomated liquid handling platforms Precision-engineered to match OEM dimensions they guarantee flawless automation performance and form a tight reliable seal with the robotic system Made from a specialized polymer that resists DNA RNA and protein adherence these tips are manufactured using diamond-polished molds to eliminate cavities or occlusions that could cause sample retention They are also free from secondary substances such as silicone ensuring sample purity Specifications Volume 200 L Color Clear Sterile Standard Style Certification RNase DNase and Pyrogen-Free Packaging 96 Tips/Rack 10 Racks/Unit 5 Units/Case