Learn More
Bellco Glass, Inc. 200 /300 Pipette Tip Rack
Supplier: Bellco Glass, Inc. 229061000
Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly. Reduce plastic waste by refilling empty racks available without a wafer to allow refill from pipette tip reload systems.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20