Learn More
Bellco Glass, Inc. 200 /300 Pipette Tip Rack
SDP
Catalog No. 502329408
Encompass_Preferred
Empty racks are available with a wafer to allow refill from bulk packed pipette tips. Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20
Catalog No. 50-232-9408 Supplier Bellco Glass, Inc. Supplier No. 229066000
Empty racks are available with a wafer to allow refill from bulk packed pipette tips. Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20