Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free 200X in DMSO) 100x1ml
SDP
Catalog No. 503107243
Encompass_Preferred
When extracting secreted proteins from culture medium in vitro protein production ceases and degradation increases Cell crude extracts contain many endogenous enzymes such as phosphatases and proteases capable of degrading proteins in the extracts The best way to improve the yield of intact proteins is to add inhibitors targeting these known enzymes The Protease Inhibitor Cocktail is used in tissue culture media to enhance protein stability It functions by inhibiting proteases responsible for the degradation of both phosphorylated and non-phosphorylated protein substrates This cocktail is provided as a supplement for tissue culture media to prevent degradation of secreted proteins This Protease Inhibitor Cocktail has been optimized and tested specifically for tissue culture media This protease inhibitor cocktail is provided in DMSO as a ready-to-use solution
Catalog No. 50-310-7243 Supplier Apexbio Technology LLC Supplier No. K1008100
When extracting secreted proteins from culture medium in vitro protein production ceases and degradation increases Cell crude extracts contain many endogenous enzymes such as phosphatases and proteases capable of degrading proteins in the extracts The best way to improve the yield of intact proteins is to add inhibitors targeting these known enzymes The Protease Inhibitor Cocktail is used in tissue culture media to enhance protein stability It functions by inhibiting proteases responsible for the degradation of both phosphorylated and non-phosphorylated protein substrates This cocktail is provided as a supplement for tissue culture media to prevent degradation of secreted proteins This Protease Inhibitor Cocktail has been optimized and tested specifically for tissue culture media This protease inhibitor cocktail is provided in DMSO as a ready-to-use solution