Learn More
VWR INTERNATIONAL MultiPlex qPCR SuperMix, 200 x 50 uL
Catalog No. NC1784193
Encompass
PerfeCTa Multiplex qPCR SuperMix, Low ROX is a 2X concentrated, ready-to-use reaction cocktail that contains all components, except primers, probe(s), and template for real-time quantitative PCR on Applied Biosystems 7500 or Stratagene MX series of real-time PCR systems.
Catalog No. NC1784193 Supplier VWR INTERNATIONAL Supplier No. 97065230
PerfeCTa Multiplex qPCR SuperMix, Low ROX is a 2X concentrated, ready-to-use reaction cocktail that contains all components, except primers, probe(s), and template for real-time quantitative PCR on Applied Biosystems 7500 or Stratagene MX series of real-time PCR systems.