Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike RBD-Fc Recombinant Protein (HPLC-verified)
Supplier:  Sino Biological 40592V02H100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (RBD) (YP_009724390.1) (Arg319-Phe541) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. NC1792206