Learn More
eMolecules​ 203854-58-4 | Fmoc-(S)-3-aminoisobutyric acid | Combi-Blocks, Inc. | MFCD03788584 | 325.364 | C19H19NO4 | 97.000 | C[C@@H](CNC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 10g | 837464676
Catalog No. 5000488631 Shop All eMolecules Inc. Products
Encompass_Preferred
Fmoc-(S)-3-aminoisobutyric acid | Combi-Blocks, Inc. | 203854-58-4 | MFCD03788584 | 325.364 | C19H19NO4 | 97.000 | C[C@@H](CNC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 10g | 837464676
Catalog No. 50-004-88631 Supplier eMolecules​ Supplier No. 837464676
Fmoc-(S)-3-aminoisobutyric acid | Combi-Blocks, Inc. | 203854-58-4 | MFCD03788584 | 325.364 | C19H19NO4 | 97.000 | C[C@@H](CNC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 10g | 837464676