Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MC-Val-Cit-PAB-Indib | 2055896-95-0 | 95.8% | 945.48 | C50H55ClN9O8 | 100 MG
SDP

Catalog No. 5000203523
Encompass_Preferred

MC-Val-Cit-PAB-Indibulin is a drug-linker conjugate that links the tubulin assembly inhibitor indibulin to a cleavable MC-Val-Cit-PAB ADC linker, intended for antibody-drug conjugate research and preclinical antitumor studies.

  • Drug-linker conjugate for ADC research
  • Payload: indibulin, a tubulin assembly inhibitor
  • Molecular weight 945.48 g/mol; formula C50H55ClN9O8
  • Purity approximately 95.8%
  • Available in small milligram quantities for preclinical work

Catalog No. 50-002-03523 Supplier Medchemexpress LLC Supplier No. HY128908100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-Val-Cit-PAB-Indibulin is a drug-linker conjugate that links the tubulin assembly inhibitor indibulin to a cleavable MC-Val-Cit-PAB ADC linker, intended for antibody-drug conjugate research and preclinical antitumor studies.

  • Drug-linker conjugate for ADC research
  • Payload: indibulin, a tubulin assembly inhibitor
  • Molecular weight 945.48 g/mol; formula C50H55ClN9O8
  • Purity approximately 95.8%
  • Available in small milligram quantities for preclinical work

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.