Learn More
Medchemexpress LLC S18-000003 | 2068119-11-7 | 99.3% | 10 MG
SDP
Catalog No. 5000234146
Encompass_Preferred
A potent, selective, orally active inhibitor of the RORγt nuclear receptor for preclinical research into Th17-mediated conditions such as psoriasis. Demonstrates sub-30 nM inhibition of human RORγt, strong selectivity versus related receptors, and documented solubility and storage properties for experimental use.
  • Potent and selective RORγt inhibition (IC50 <30 nM).
  • Selective over other ROR family members (IC50 >10 μM).
  • Orally active, enabling in vivo study applications.
  • High purity for reproducible experimental results.
  • Solid white to yellow form for straightforward handling.
  • High solubility in DMSO (100 mg/mL) for stock preparation.
  • Stable as powder at -20°C for long-term storage.
Catalog No. 50-002-34146 Supplier Medchemexpress LLC Supplier No. HY11936610MG
A potent, selective, orally active inhibitor of the RORγt nuclear receptor for preclinical research into Th17-mediated conditions such as psoriasis. Demonstrates sub-30 nM inhibition of human RORγt, strong selectivity versus related receptors, and documented solubility and storage properties for experimental use.
  • Potent and selective RORγt inhibition (IC50 <30 nM).
  • Selective over other ROR family members (IC50 >10 μM).
  • Orally active, enabling in vivo study applications.
  • High purity for reproducible experimental results.
  • Solid white to yellow form for straightforward handling.
  • High solubility in DMSO (100 mg/mL) for stock preparation.
  • Stable as powder at -20°C for long-term storage.