Learn More
Chemglass Life Sciences Flange, Compatible with Buchi Rotary Evaporators, Flange OD: 210mm, Flange ID: 176mm, Shank OD: 190mm
SDP
Catalog No. CG14606
Encompass_Preferred
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 210mm, Flange ID: 176mm, Shank OD: 190mm.
Catalog No. CG14606 Supplier Chemglass Life Sciences Supplier No. CG14606
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 210mm, Flange ID: 176mm, Shank OD: 190mm.