Learn More
MSC Hubbell Wiring Device-Kellems HBL7411C 120/208 VAC 20A NonNEMA Industrial Twist Lock Plug 4 Poles, 4 Wires, 3 Phase, IP20, Ungrounded, 0.35 to 1.15" Cord Diam, Black/White
Catalog No. 501297239
Encompass
Hubbell Wiring Device-Kellems HBL7411C 120/208 VAC 20A NonNEMA Industrial Twist Lock Plug 4 Poles, 4 Wires, 3 Phase, IP20, Ungrounded, 0.35 to 1.15" Cord Diam, Black/White
Catalog No. 50-129-7239 Supplier MSC Supplier No. 54035266
Hubbell Wiring Device-Kellems HBL7411C 120/208 VAC 20A NonNEMA Industrial Twist Lock Plug 4 Poles, 4 Wires, 3 Phase, IP20, Ungrounded, 0.35 to 1.15" Cord Diam, Black/White