Learn More
MSC Ferraz Shawmut TRS20R 300 VDC, 600 VAC, 20 Amp, Time Delay General Purpose Fuse Clip Mount, 127mm Overall Length, 20 at DC, 200 at AC kA Rating, 13/16" Diam
Supplier:  MSC 54006440
Encompass
Ferraz Shawmut TRS20R 300 VDC, 600 VAC, 20 Amp, Time Delay General Purpose Fuse Clip Mount, 127mm Overall Length, 20 at DC, 200 at AC kA Rating, 13/16" Diam
Catalog No. 50-129-7261