Learn More
Cayman Chemical 20a-dIhydro PrednIsolon 10mg
SDP
Catalog No. 502935459
Encompass_Preferred
A metabolite of prednisolone; has been found in the urine of cattle following ACTH administration but not in naturally stressed or non-stressed cattle
Catalog No. 50-293-5459 Supplier Cayman Chemical Supplier No. 2650210 MG
A metabolite of prednisolone; has been found in the urine of cattle following ACTH administration but not in naturally stressed or non-stressed cattle