Learn More
Cayman Chemical 20carboxy Arachidonic Acid
SDP
Catalog No. 502890994
Encompass_Preferred
The major metabolite of 20-HETE that is produced in renal tubular epithelial, endothelial, and microvascular smooth muscle cell cultures; ω-oxidation conversion can take place using purified alcohol dehydrogenases three and four or by microsomes containing recombinant human CYP4F3B; 20-carboxy arachidonic acid inhibits ion transport in the kidneys; produces vasorelaxation of porcine coronary microvessels constricted with endothelin; binds to isolated ligand binding domains of peroxisome proliferator-activated receptor α (PPARα) (Kd = 0.87 ± 0.12 µM) and PPARγ (Kd = 1.7 ± 0.5 µM); a dual activator of PPARα and PPARγ in a transiently transfected COS-7 cell reporter system
Catalog No. 50-289-0994 Supplier Cayman Chemical Supplier No. 1000791225 UG
Add to Cart
Add to Cart
The major metabolite of 20-HETE that is produced in renal tubular epithelial, endothelial, and microvascular smooth muscle cell cultures; ω-oxidation conversion can take place using purified alcohol dehydrogenases three and four or by microsomes containing recombinant human CYP4F3B; 20-carboxy arachidonic acid inhibits ion transport in the kidneys; produces vasorelaxation of porcine coronary microvessels constricted with endothelin; binds to isolated ligand binding domains of peroxisome proliferator-activated receptor α (PPARα) (Kd = 0.87 ± 0.12 µM) and PPARγ (Kd = 1.7 ± 0.5 µM); a dual activator of PPARα and PPARγ in a transiently transfected COS-7 cell reporter system