Learn More
Alkali Scientific 20' Cassette of 12mm ULT (Ultra-low Temp) Lab Tape, White with Black Print
SDP
Catalog No. 502336045
Encompass_Preferred
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This white tape with black print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50˚C. Special Ultra-Low Temp label tapes are also available for storage in -80˚C environments or for liquid nitrogen applications.
Catalog No. 50-233-6045 Supplier Alkali Scientific Supplier No. L901012ULT
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This white tape with black print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50˚C. Special Ultra-Low Temp label tapes are also available for storage in -80˚C environments or for liquid nitrogen applications.