Learn More
Terumo Medical Corporation 20cc Syringe with Eccentric Luer Tip
Catalog No. 23600038 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Volume (Metric):
20 mL
Catalog No. Volume (Metric)
23-600-038 20 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-600-038 Supplier Terumo Medical Corporation Supplier No. SS20ES2
Add to Cart
Add to Cart
This item is not returnable. View return policy
Graduated syringe without needle
  • Eccentric luer tip
  • Graduations: 1cc (25cc total)
  • 50 / box, 4 boxes / case (200)
Specifications
Material Polypropylene (Barrel), Polypropylene (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe without needle
Disposable Single-use
Graduations 1 cc
Volume (Metric) 20 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Eccentric Luer Tip
Graduated Yes
Sterility Sterile