Learn More
Horizon Scientifc Manual Defrost Laboratory Freezer with Hydrocarbon Refrigerants
SDP
Catalog No. NC1868148
Encompass_Preferred
These manual defrost laboratory freezers offer professional cold storage for scientific and clinical settings. Units come equipped with temperature alarms, remote alarm contacts, keyed door locks, probe access, and precise temperature monitoring through microprocessor control. These freezers provide multi-functional features in a cost-effective design. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
These products are UL/cUL Listed and come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Catalog No. NC1868148 Supplier Horizon Scientifc Supplier No. NSPF201WWW/0MHC
These manual defrost laboratory freezers offer professional cold storage for scientific and clinical settings. Units come equipped with temperature alarms, remote alarm contacts, keyed door locks, probe access, and precise temperature monitoring through microprocessor control. These freezers provide multi-functional features in a cost-effective design. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
These products are UL/cUL Listed and come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.