Learn More
Braintree Scientific Malleable S/S Disposable Feeding Needle, Sterile, 20G-1.5""
SDP
Catalog No. NC9079385
Encompass
Malleable S/S Disposable Feeding Needle. These needles feature plastic luer hubs and silicone tips and can be easily formed to desired shape. 100 sterile needles per box. **Sold for research purposes only and not for human use**
Catalog No. NC9079385 Supplier Braintree Scientific Supplier No. D001
Malleable S/S Disposable Feeding Needle. These needles feature plastic luer hubs and silicone tips and can be easily formed to desired shape. 100 sterile needles per box. **Sold for research purposes only and not for human use**