Learn More
Ace Glass, Inc. 20g 152mm stainless steel needles, cs/12, sp/6
SDP
Catalog No. 509559110
Encompass
20 gauge 152mm stainless steel needle with a deflected septum point and a standard female hub. Case of 12.
Catalog No. 50-955-9110 Supplier Ace Glass, Inc. Supplier No. 1368407
20 gauge 152mm stainless steel needle with a deflected septum point and a standard female hub. Case of 12.