Learn More
Ace Glass, Inc. 20g 305mm stainless steel needles, cs/12, sp/6
SDP
Catalog No. NC0765411
Encompass
20 gauge 305mm stainless steel needle with a deflected septum point and a standard female hub. Case of 12.
Catalog No. NC0765411 Supplier Ace Glass, Inc. Supplier No. 1368411
20 gauge 305mm stainless steel needle with a deflected septum point and a standard female hub. Case of 12.