Learn More
MSC Loctite 135429 0.70 oz Bottle Clear Instant Adhesive Series 401, 15 sec Fixture Time, 24 hr Full Cure Time, Bonds to Plastic & Rubber
Catalog No. 501113862
Encompass
Loctite 135429 0.70 oz Bottle Clear Instant Adhesive Series 401, 15 sec Fixture Time, 24 hr Full Cure Time, Bonds to Plastic & Rubber
Catalog No. 50-111-3862 Supplier MSC Supplier No. 88545264
Loctite 135429 0.70 oz Bottle Clear Instant Adhesive Series 401, 15 sec Fixture Time, 24 hr Full Cure Time, Bonds to Plastic & Rubber