Learn More
MSC Loctite Instant Adhesive Gel, Clear, High Viscosity, 20 g
Catalog No. 501113863
Encompass
LOCTITE 454 is a clear, gel viscosity instant adhesive formulated for rapid bonding across a wide range of materials, including metals, elastomers, and plastics. It is also well-suited for porous and absorbent substrates like wood, paper, and fabrics, making it ideal for large bond gaps and vertical applications.
  • Gel viscosity for drip-free overhead and vertical applications
  • Rapid bonding to metals, elastomers, plastics, and porous materials
  • Operates across a wide temperature range, -65°F to 250°F
  • Achieves fast fixture strength
  • Provides high tensile strength
Catalog No. 50-111-3863 Supplier MSC Supplier No. 88545280
LOCTITE 454 is a clear, gel viscosity instant adhesive formulated for rapid bonding across a wide range of materials, including metals, elastomers, and plastics. It is also well-suited for porous and absorbent substrates like wood, paper, and fabrics, making it ideal for large bond gaps and vertical applications.
  • Gel viscosity for drip-free overhead and vertical applications
  • Rapid bonding to metals, elastomers, plastics, and porous materials
  • Operates across a wide temperature range, -65°F to 250°F
  • Achieves fast fixture strength
  • Provides high tensile strength