Learn More
Cml Supply 20G X 1-1/2IN BLUNT 1000PK
Catalog No. NC9290189
Encompass_Preferred
Replacement Needles; Blunt Tip Industrial Dispensing Needle; Brico Medical Supplies; Stainless steel; 20g x 1-1/2 In. Luer Lock Polypropylene Hub; Autoclavable; 1000/Pk
Catalog No. NC9290189 Supplier Cml Supply Supplier No. 90120150M
Replacement Needles; Blunt Tip Industrial Dispensing Needle; Brico Medical Supplies; Stainless steel; 20g x 1-1/2 In. Luer Lock Polypropylene Hub; Autoclavable; 1000/Pk