Learn More
MSC Securall Cabinets Flammable Liquid Cabinet, Manual Close, 20 gal
Catalog No. 502886331
Encompass
This Securall steel cabinet features a manual closing door, two shelves, and a yellow finish. Measuring 44 in. high, 18 in. deep, and 24 in. wide, it meets NFPA Code 30 and OSHA standards.
  • Double-wall construction (18 gauge) with 1-1/2 in. air space and seamed joints
  • Deep, 2 in. leak-proof sill
  • Two 2 in. vents with flash arrestors
  • Non-sparking design
  • Adjustable leveling legs and static grounding connection
  • Full length, heavy-duty piano hinges and flush mounted locking handle
  • Manual closing door type
  • Suitable for flammable and combustible liquids
  • Three-point key lock type
  • Complies with NFPA Code 30 and OSHA standards
  • FM approved
Catalog No. 50-288-6331 Supplier MSC Supplier No. 55007587
This Securall steel cabinet features a manual closing door, two shelves, and a yellow finish. Measuring 44 in. high, 18 in. deep, and 24 in. wide, it meets NFPA Code 30 and OSHA standards.
  • Double-wall construction (18 gauge) with 1-1/2 in. air space and seamed joints
  • Deep, 2 in. leak-proof sill
  • Two 2 in. vents with flash arrestors
  • Non-sparking design
  • Adjustable leveling legs and static grounding connection
  • Full length, heavy-duty piano hinges and flush mounted locking handle
  • Manual closing door type
  • Suitable for flammable and combustible liquids
  • Three-point key lock type
  • Complies with NFPA Code 30 and OSHA standards
  • FM approved