Learn More
GLOMED INC Reusable Animal Feeding Needle 20G x 38mm Straight
Catalog No. NC1299558
Encompass_Preferred
Reusable Metal Hub with luer lock fitting - Smooth stainless steel ball tip and stainless steel tubing - Can be used as perfusion needle - Packed individually - Sold unsterile - please sterilize before use
This item is not returnable. View return policy
Catalog No. NC1299558 Supplier GLOMED INC Supplier No. AFN2038S
Reusable Metal Hub with luer lock fitting - Smooth stainless steel ball tip and stainless steel tubing - Can be used as perfusion needle - Packed individually - Sold unsterile - please sterilize before use